Learn More
Thermo Scientific™ RIPA Lysis and Extraction Buffer
Lyse cultured mammalian cells with this high-quality, ready-to-use and fully disclosed formulation of a popular cell lysis reagent.
£89.00 - £193.00
Specifications
Content And Storage | Upon receipt store at 4°C. |
---|---|
Format | Liquid |
Product Type | Extraction Buffer |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10017003
|
Thermo Scientific™
89900 |
100 mL |
£89.00
100mL |
In Stock
Log in to see stock. |
|||||
10230544
|
Thermo Scientific™
89901 |
250 mL |
£193.00
250mL |
In Stock
Log in to see stock. |
|||||
Description
Thermo Scientific™ RIPA Buffer is a high-quality, ready-to-use and fully disclosed formulation of a popular cell lysis reagent for cultured mammalian cells.
This RIPA buffer effectively lyses and extracts protein from cultured mammalian cells, including plated cells and pelleted suspension cells. The popular reagent enables the extraction of membrane, nuclear and cytoplasmic proteins and is compatible with many applications, including reporter assays, the Thermo Scientific BCA Protein Assay, immunoassays and protein purification. Inhibitors such as Thermo Scientific Halt Protease Inhibitor Cocktail (Part No. 78430) and Halt Phosphatase Inhibitor Cocktail (Part No. 78420) are also compatible with this RIPA buffer formulation and can be added before use to prevent proteolysis and maintain protein phosphorylation.Highlights:
Convenient – ready-to-use solution
Flexible – compatible with many applications, including reporter assays, protein assays, immunoassays and protein purification
Versatile – enables extraction of cytoplasmic, membrane and nuclear proteins
Disclosed formulation – contains no proprietary components, providing users with complete control and knowledge of possible compatibility issues
Specifications
Upon receipt store at 4°C. | |
Liquid | |
Extraction Buffer |
For Research Use Only. Not for use in diagnostic procedures.