Learn More
Thermo Scientific™ Pierce™ SDS-PAGE Sample Prep Kit
Quickly and easily remove high levels of salts, denaturants, detergents and other buffer components that interfere with polyacrylamide gel electrophoresis.
Brand: Thermo Scientific™ 89888
185.74 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : PN
Additional Details : Weight : 2.72160kg
Description
Thermo Scientific™ Pierce SDS-PAGE Sample Prep Kit quickly and easily removes high levels of salts, denaturants, detergents and other buffer components that interfere with polyacrylamide gel electrophoresis of proteins.
The SDS-PAGE Sample Prep Kit provides a simple and effective method for concentrating samples while removing chemicals that interfere with SDS-PAGE analysis, such as acids and bases, detergents, guanidine and ammonium sulfate. The proprietary PAGE-prep Resin binds protein in the presence of dimethylsulfoxide (DMSO). Interfering contaminants are then removed by washing the protein-bound resin in an easy-to-use spin cup format. Proteins are eluted in a buffer compatible with the Pierce BCA Protein Assay, allowing quantitation of a portion of the processed sample. The eluted proteins can then be combined with sample buffer and analyzed on the SDS-PAGE system of choice.Highlights:
Eliminates gel artifacts caused by incompatible contaminants – removes dyes, reducing agents, detergents, sugars, glycerol, guanidine, urea and ammonium sulfate to provide reproducible results by SDS-PAGE analysis
Compatible with the BCA protein assay – allows protein quantitation of the processed sample before gel-loading
Enriches dilute protein solutions – concentrates protein samples for SDS-PAGE analysis by six-fold in less than 20 minutes
Powerful and flexible – process 2 to 350μL of sample containing up to 70μg of protein; recover 75 to 85% of the total protein in 50μL of elution buffer
Convenient, complete kit – includes PAGEprep Resin, DMSO, spin cups and sample loading buffer
-
Includes:
PAGEprep resin slurry, DMSO solution, elution buffer, sample buffer, spin cups and collection tubes
-
Specifications
Upon receipt store kit at room temperature. | |
SDS-PAGE | |
SDS-PAGE Gels |
Sample Loading Buffers | |
SDS-PAGE Sample Prep Kit | |
Pierce™ |
Safety and Handling
- Pierce SDS-PAGE Sample Prep Kit
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H351-Suspected of causing cancer.
- H373-May cause damage to organs.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P314-Get medical advice/attention if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Kieselguhr, calcined
For Research Use Only. Not for use in diagnostic procedures.