Learn More
Thermo Scientific™ SIA (succinimidyl iodoacetate)
Crosslink amines to sulfhydryls with this very short crosslinker composed of NHS-ester and iodoacetyl (haloacetyl) groups at ends of a 3-atom (1.5A) spacer arm.
Brand: Thermo Scientific™ 22349
Description
Thermo Scientific™ Pierce SIA is a very short (1.5 Å) crosslinker for amine-to-sulfhydryl conjugation via N-hydroxysuccinimide (NHS) ester and iodoacetyl reactive groups.
Suited to capturing amine and sulfhydryl groups in very close proximityHighlights:
- Reactive groups: NHS ester and iodoacetyl
- Reactive toward: amino and sulfhydryl groups
Specifications
Amine-Sulfhydryl | |
SIA | |
No | |
Upon receipt store at 4°C protected from moisture. | |
Ambient | |
Chemical Labeling | |
Haloacetyl, NHS Ester | |
Solid | |
DMF, DMSO | |
No |
No | |
283.02 | |
1.5 Å | |
Yes | |
Pierce™ | |
Heterobifunctional | |
Short (<10 Å) | |
50 mg | |
Standard | |
Crosslinker |
Safety and Handling
- SIA (succinimidyl iodoacetate)
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : N-Hydroxysuccinimidyl iodoacetate
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.