missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Powdered DMEM Medium for SILAC

Analyze protein expression by mass spectrometry using these media or additives for stable isotope labeling with amino acids in cell culture (SILAC).

Brand:  Thermo Scientific™ 88425

Product Code. 12172410

  • £350.00 / 135g
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

These Thermo Scientific Pierce cell culture media and serum components are optimized for use with stable isotope labeling with amino acids in cell culture (SILAC) to analyze protein expression by mass spectrometry (MS).

SILAC is a powerful method to identify and quantify relative differential changes in complex protein samples. This approach entails the in vivo metabolic incorporation of heavy 13C- or 15N-labeled amino acids into proteins followed by mass spectrometry (MS) for the accelerated and comprehensive identification, characterization and quantitation of proteins.

Highlights:

  • Flexible–liquid media deficient in both L-lysine and L-arginine and powdered media deficient in L-leucine, L-lysine and L-arginine, allowing for more complete proteome coverage through multiple isotopic amino acid labeling

  • Compatible–DMEM, MEM, RPMI-1640, IMDM, Ham's F12 and McCoy's 5A media and dialyzed FBS are used to culture a variety of cell lines, including HeLa, 293T, COS7, U2OS, A549, A431, HepG2, NIH 3T3, K562 and Jurkat

  • Optimized, specialized media–the Expansion DMEM and Low-Osmolarity DMEM are specifically designed and qualified for mouse embryonic stem cell propagation and SILAC protein expression profiling with mass spectrometry

  • High quality–Stem Cell Screened Dialyzed FBS is tested to ensure that it does not promote stem cell differentiation

TRUSTED_SUSTAINABILITY
Specifications

Specifications

DMEM media minus L-Leucine, L-Lysine and L-Arginine
Media
Wet Ice
Protein
135 g
Mass Spectrometer
135 g of powdered medium to make 10 L of medium.

Store in refrigerator (2 to 8°C).
Cells
Protein Labeling
Powder
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.