Learn More
Thermo Scientific™ McCoy's 5A Media for SILAC
Analyze protein expression by mass spectrometry using these media or additives for stable isotope labeling with amino acids in cell culture (SILAC).
Brand: Thermo Scientific™ 88441
Code : A0
Additional Details : Weight : 0.01000kg
Description
These Thermo Scientific Pierce cell culture media and serum components are optimized for use with stable isotope labeling with amino acids in cell culture (SILAC) to analyze protein expression by mass spectrometry (MS).
SILAC is a powerful method to identify and quantify relative differential changes in complex protein samples. This approach entails the in vivo metabolic incorporation of heavy 13C- or 15N-labeled amino acids into proteins followed by mass spectrometry (MS) for the accelerated and comprehensive identification, characterization and quantitation of proteins.Highlights:
Flexible–liquid media deficient in both L-lysine and L-arginine and powdered media deficient in L-leucine, L-lysine and L-arginine, allowing for more complete proteome coverage through multiple isotopic amino acid labeling
Compatible–DMEM, MEM, RPMI-1640, IMDM, Ham's F12 and McCoy's 5A media and dialyzed FBS are used to culture a variety of cell lines, including HeLa, 293T, COS7, U2OS, A549, A431, HepG2, NIH 3T3, K562 and Jurkat
Optimized, specialized media–the Expansion DMEM and Low-Osmolarity DMEM are specifically designed and qualified for mouse embryonic stem cell propagation and SILAC protein expression profiling with mass spectrometry
High quality–Stem Cell Screened Dialyzed FBS is tested to ensure that it does not promote stem cell differentiation
Specifications
McCoy's 5A media minus L-Lysine and L-Arginine | |
Media | |
Wet Ice | |
Protein | |
500 mL |
Mass Spectrometer | |
Store in refrigerator (2 to 8°C). | |
Cells | |
Protein Labeling | |
Liquid |
For Research Use Only. Not for use in diagnostic procedures.