missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ SMCC

Product Code. 10066813 Shop All Thermo Scientific Products
Click to view available options
Quantity:
50 mg
10 x 1 mg
Unit Size:
50mg
Pack of 10
This item is not returnable. View return policy

Product Code. 10066813

Brand: Thermo Scientific™ 22360

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

No-Weigh™ Format

Thermo Scientific™ Pierce SMCC is an amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a medium-length cyclohexane-stabilized spacer arm (8.3 Ås).

Highlights:

  • Cyclohexane bridge adds stability to the maleimide-reactive group; N-(4-carboxycyclohexylmethyl) maleimide groups are stable for 64 hours in 0.1 M sodium phosphate buffer, pH 7.0 at 4°C
  • Ideal for coupling enzymes to antibodies
  • Reactive groups: NHS-ester and maleimide
  • Reactive toward: amino and sulfhydryl groups
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine-Sulfhydryl
Cleavable By Thiols
Molecular Weight (g/mol) 334.32
PEGylated No
Spacer Arm Length 8.3 Å
Content And Storage Upon receipt store desiccated at 4°C.
Cell Permeability Yes
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Maleimide, NHS Ester
Spacer Short (<10 Å)
Form Powder
Quantity 50 mg
Solubility DMF, DMSO
Format Standard, Single-use
Water Soluble No
Product Type Crosslinker
Show More Show Less
compliance-icons
Product Identifier
  • SMCC
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Succinimidyl-trans-4-(N-maleimidylmethyl)cyclohexane-1-carboxylate

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.