Learn More
Thermo Scientific™ Sodium Acetate (3 M)
Consitently purify and store DNA without any worries. Thermo Scientific™ Pierce™ Sodium Acetate Solution (3M) is a highly purified, sterile 3M sodium acetate solution.
Brand: Thermo Scientific™ 17888
Code : PN
Additional Details : CAS Number : 6131-90-4 Weight : 1.00000kg
Description
Thermo Scientific™ Pierce Sodium Acetate Solution is a sterile, 3M soluion of sodium acetate that is useful for DNA purification and storage.
Additional Information and Online Ordering »
Thermo Scientific™ Pierce™ Sodium Acetate Solution is a sterile, 3M solution of sodium acetate that is useful for DNA purification and storage. The clear, colorless and odorless liquid is delivered in 1L bottles.
Specifications
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25°C. | |
Sodium Acetate Solution | |
1 L | |
Liquid |