missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Stop Solution for TMB Substrates

Stop TMB-based ELISA detection with this 0.16M sulfuric acid for use with the ELISA substrate 3,3',5,5' - tetramethylbenzidine (TMB).

Brand:  Thermo Scientific™ N600

93.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.



Product Code. 10187433

  • £101.00 / 55mL
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce Stop Solution for TMB is 0.16M sulfuric acid for use with the ELISA substrate 3,3',5,5' - tetramethylbenzidine (TMB).

TMB Stop Solution is 0.16M sulfuric acid for use with the ELISA substrate 3,3«,5,5«-tetramethylbenzidine (TMB). Addition of sulfuric acid stop solution changes the color from blue to yellow, stabilizing the color development to enable accurate measurement of the intensity at 450nm using a spectrophotometer or plate reader.
  • Size: 55mL

  • Sufficient For: five 96-well microplates

  • Application: ELISA

Instructions: Following TMB incubation, add an amount of stop solution equal to the amount of TMB substrate added to each well of the ELISA plate (typically 50 to 100μL per well). Use at full strength.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Stop Solution
Liquid
Store at 2–8°C.
55 mL
ELISA
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Stop Solution for TMB Substrates > 55mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.