missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Streptavidin

Product Code. 10147723 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 mg
5 mg
100 mg
1000 mg
Unit Size:
1000mg
100mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 10147723

Brand: Thermo Scientific™ 21122

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Purified recombinant streptavidin enables detection of biotinylated antibodies and other probes in a variety of standard assay methods including western blotting, ELISA, immunohistochemistry, and fluorescence imaging.

  • Purified streptavidin is supplied lyophilized in phosphate-based buffer for immediate reconstitution with water
  • Streptavidin is a biotin-binding protein originally isolated from Streptomyces avidinii
  • Pierce™ streptavidin products use a recombinant form of streptavidin having a mass of 53,000 daltons and a near-neutral pI
  • Streptavidin is a tetrameric protein, with each subunit binding one molecule of biotin with affinity similar to that of avidin
  • Guanidinium chloride will dissociate avidin and streptavidin into subunits, but streptavidin is more resistant to dissociation
  • Applications: ELISA (assay dependent), western blot (assay dependent)
TRUSTED_SUSTAINABILITY

Specifications

Conjugate Unconjugated
Quantity 1 mg
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C.
Form Solid
Product Line Pierce
Product Type Streptavidin
Target Molecule Biotin
Label or Dye Unlabeled

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.