missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Streptavidin Coated High Capacity Plates

Thermo Scientific™ Pierce Streptavidin High Binding Capacity (HBC) Coated Plates are designed for binding biotinylated oligonucleotides and peptides with higher binding efficiency than other commercially available plates.

Brand:  Thermo Scientific™ 15502

247.95 GBP valid until 2024-11-30
Use promo code "21681" to get your promotional price.


 View more versions of this product

Product Code. 10003413

  • £261.00 / Pack of 5
Estimated Shipment: 22-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Pierce Streptavidin High Binding Capacity (HBC) Plates are prepared with an exclusive coating method that yields a streptavidin-coated plate with four to five times the binding capacity of other commercially available polystyrene assay plates. Using a Pierce Streptavidin HBC Plate can result in an assay with a broader dynamic range and better linearity, leading to improved assay precision. The plates are offered in several plate formats, plate colors, and coating volumes.

Thermo Scientific Pierce Streptavidin-Coated High-Capacity Plates are designed for the binding of biotinylated oligonucleotides and peptides with higher binding efficiency than other commercially available plates.

Pierce Streptavidin-coated plates are widely used for applications such as ELISAs and other assay protocols. These biotin-binding plates come with different capacities and sensitivities for maximum flexibility. For example, the high-binding-capacity plates (HBC) can be used for anti-therapeutic/anti-drug antibodies (ADA) screening for immunogenicity research and biologics development (1,2). Both regular and HBC plates have been used in aptamer assays for clinical and industrial needs, along with neurobiology research for therapeutics and prevention, amongst other applications (3–5).

Features of Streptavidin-Coated High-Capacity Plates include:
• Unique plate-coating technology—results in high loading and binding capacity
• Streptavidin-coated option—plates have up to five times the binding capacity of typical plates
• Broader dynamic range—extends the quantitative range so there is less need for dilutions
• Save time—pre-blocked plates to reduce the number of assay steps
• Flexible assay formats—coated plates offered in 8-well strips to reduce waste

The proprietary plate-coating technology used to manufacture these Pierce coated plates yields high-capacity plates with a wider detection limit than regular-binding-capacity plates. The standard curve exhibits greater linearity for detecting small biotinylated molecules such as peptides and oligonucleotides, resulting in greater assay precision.

References
1. J Immunol Methods. 2016 Apr;431:45-51
2. J Immunol Methods. 2011 Feb 28;365(1-2):38-49
3. Cancers (Basel). 2022 Oct 25;14(21):5222
4. J Microbiol Biotechnol. 2013 Jun 28;23(6):878-84
5. Acta Neuropathol Commun. 2018 May 31;6(1):43

TRUSTED_SUSTAINABILITY
Specifications

Specifications

High Capacity, Affinity
White, 96-well
Pierce
Microplate
96
5 Plates
Upon receipt store plates at 4°C in unopened pouches. Once opened, place unused plates in a resealable bag with desiccant and store at 4°C.
Chemiluminescence
White
Polystyrene
Streptavidin
SuperBlock™ Buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers