missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Streptavidin Coated High Capacity Plates

Product Code. p-7227875 Shop All Thermo Scientific Products
Click to view available options
Color:
Black
Clear
White
No. of Wells:
96
384
Format:
Standard
Strip Plate
Detection Method:
Chemiluminescence
Colorimetric
Fluorescence
Unit Size:
Pack of 5
This item is not returnable. View return policy

Product Code. 10532964

Brand: Thermo Scientific™ 15503

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ Pierce Streptavidin High Binding Capacity (HBC) Coated Plates are designed for binding biotinylated oligonucleotides and peptides with higher binding efficiency than other commercially available plates.

Pierce Streptavidin High Binding Capacity (HBC) Plates are prepared with an exclusive coating method that yields a streptavidin-coated plate with four to five times the binding capacity of other commercially available polystyrene assay plates. Using a Pierce Streptavidin HBC Plate can result in an assay with a broader dynamic range and better linearity, leading to improved assay precision. The plates are offered in several plate formats, plate colors, and coating volumes.

Thermo Scientific Pierce Streptavidin-Coated High-Capacity Plates are designed for the binding of biotinylated oligonucleotides and peptides with higher binding efficiency than other commercially available plates.

Pierce Streptavidin-coated plates are widely used for applications such as ELISAs and other assay protocols. These biotin-binding plates come with different capacities and sensitivities for maximum flexibility. For example, the high-binding-capacity plates (HBC) can be used for anti-therapeutic/anti-drug antibodies (ADA) screening for immunogenicity research and biologics development (1,2). Both regular and HBC plates have been used in aptamer assays for clinical and industrial needs, along with neurobiology research for therapeutics and prevention, amongst other applications (3–5).

Features of Streptavidin-Coated High-Capacity Plates include:
• Unique plate-coating technology—results in high loading and binding capacity
• Streptavidin-coated option—plates have up to five times the binding capacity of typical plates
• Broader dynamic range—extends the quantitative range so there is less need for dilutions
• Save time—pre-blocked plates to reduce the number of assay steps
• Flexible assay formats—coated plates offered in 8-well strips to reduce waste

The proprietary plate-coating technology used to manufacture these Pierce coated plates yields high-capacity plates with a wider detection limit than regular-binding-capacity plates. The standard curve exhibits greater linearity for detecting small biotinylated molecules such as peptides and oligonucleotides, resulting in greater assay precision.

References
1. J Immunol Methods. 2016 Apr;431:45-51
2. J Immunol Methods. 2011 Feb 28;365(1-2):38-49
3. Cancers (Basel). 2022 Oct 25;14(21):5222
4. J Microbiol Biotechnol. 2013 Jun 28;23(6):878-84
5. Acta Neuropathol Commun. 2018 May 31;6(1):43

TRUSTED_SUSTAINABILITY

Specifications

Binding Property High Capacity, Affinity
Format Standard
Content And Storage Upon receipt store plates at 4°C in unopened pouches. Once opened, place unused plates in a resealable bag with desiccant and store at 4°C.
Detection Method Fluorescence
Product Line Pierce
Color Black
Product Type Microplate
Material Polystyrene
No. of Wells 96
Surface Treatment Streptavidin
No. per Pack 5 Plates
Plate Blocking SuperBlock™ Buffer
Quantity 5 plates
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.