missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ High Sensitivity Streptavidin-HRP, Pre-Diluted

Product Code. 11806824 Shop All Thermo Scientific Products
Click to view available options
Quantity:
2 mL
Unit Size:
2mL
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 11806824

missing translation for 'mfr': Thermo Scientific™ 21134

Please to purchase this item. Need a web account? Register with us today!

Edge
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Amplify signal in Western blot, ELISA or other biotin-based detection methods with this exclusive, high signal-to-noise, peroxidase-conjugated, ready-to-use streptavidin.

Thermo Scientific™ Pierce High Sensitivity Streptavidin HRP Conjugate is an exclusive peroxidase-conjugated streptavidin biotin-binding protein that provides signal amplification like poly-HRP and exceptional storage stability.

This specially-manufactured variety of HRP-conjugated streptavidin protein meets the demands of today's scientists for more sensitive detection in ELISA and Western blotting applications. The conjugate is suitable for use with chemiluminescent, chemifluorescent or colorimetric substrates. Each High Sensitivity HRP conjugate is packaged in an easy-to-use stabilized solution that enables convenient storage at 4°C for at least one year.

Highlights:

  • High sensitivity – detect low levels of target without background; obtain high signal-to-noise ratios

  • Cost effective – use less conjugate in Western blotting and ELISA applications and still obtain excellent results

  • Flexible – compatible with typical chemiluminescent, fluorescent and colorimetric peroxidase substrates

  • Convenient – ready-to-use stabilized liquid format means there is no waiting for thawing and no need to aliquot

TRUSTED_SUSTAINABILITY

Spezifikation

Quantity 2 mL
Target Molecule Biotin
Content And Storage Storage at 4°C for at least one year.
Specificity Biotin
Detection Method Colorimetric, Chemiluminescence, Chemifluorescence
Enzyme HRP (Horseradish Peroxidase)
Form Liquid
For Use With (Application) Western Blotting, ELISA
Product Line Pierce
Label or Dye HRP (Horseradish Peroxidase)
compliance-icons
Produktidentifikator
  • Pierce High Sensitivity Streptavidin-HRP, Pre-Diluted
Signalwort
  • Warning
Gefahrenkategorie
  • Skin sensitiser Category 1
Gefahrenhinweis
  • H317-May cause an allergic skin reaction.
Sicherheitshinweis
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Ergänzende Informationen
  • MIXTURE LIST-Contains : 5-chloro-2-methyl-4-isothiazolin-3-one,2-methyl-2H-isothiazol-3-one

For Research Use Only. Not for use in diagnostic procedures.

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt