missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ High Sensitivity Streptavidin-HRP

Amplify signal in Western blot, ELISA or other biotin-based detection methods with this exclusive, high signal-to-noise, peroxidase-conjugated, ready-to-use streptavidin.

Brand:  Thermo Scientific™ 21132

 View more versions of this product

Product Code. 11896814

  • £988.00 / 5mL
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce High Sensitivity Streptavidin HRP Conjugate is an exclusive peroxidase-conjugated streptavidin biotin-binding protein that provides signal amplification like poly-HRP and exceptional storage stability.

  • Thermo Scientific Pierce High Sensitivity Streptavidin-HRP is an exclusive peroxidase-conjugated streptavidin biotin-binding protein
  • It provides signal amplification like poly-HRP and exceptional storage stability
  • This specially-manufactured variety of HRP-conjugated streptavidin protein meets the demands of today's scientists for more sensitive detection in ELISA and Western blotting applications
  • Conjugate is suitable for use with chemiluminescent, chemifluorescent, or colorimetric substrates
  • Each High Sensitivity HRP conjugate is packaged in an easy-to-use stabilized solution that enables convenient storage at 4°C for at least one year
  • Experiments indicate that the High Sensitivity Streptavidin-HRP can be diluted 8-to 10-fold more than traditional HRP conjugates and still produce similar results
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1.0–1.1 mg/mL
5 mL
Pierce™
HRP
Storage at 4°C for at least one year.
Biotin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.