missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Streptavidin Protein, AP

Streptavidin Protein, AP conjugate, 21324, from Invitrogen™, 21324, from Invitrogen™ Species Reactivity: ; Applications: ELISA, Western Blot.

Brand:  Thermo Scientific™ 21324

266.95 GBP valid until 2024-12-31
Use promo code "21979" to get your promotional price.


Product Code. 10531474

  • £281.00 / 1mg
Estimated Shipment: 29-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Streptavidin Protein, AP conjugate, 21324, from Invitrogen., 21324, from Invitrogen. Species Reactivity: ; Applications: ELISA, Western Blot.

Streptavidin is biotin-binding protein that was originally isolated from Streptomyces avidinii. In contrast to avidin, streptavidin has no carbohydrate and has a mildly acidic pI of 5. Invitrogen Streptavidin products use a recombinant form of streptavidin having a mass of 53,000 daltons and a near-neutral pI. Streptavidin is a tetrameric protein, with each subunit binding one molecule of biotin with affinity similar to that of avidin. Guanidinium chloride will dissociate avidin and streptavidin into subunits, but streptavidin is more resistant to dissociation.
Formulation: 1mg purified protein (2-4mg/mL) in TBS with 50% glycerol, 0.1mMmgCl2, 0.1mM ZnCl2 and no preservative; pH 8
This product has been successfully used in ELISA and Western Blot procedures.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.1 μg/mL
1 mg
Liquid
Streptavidin Conjugate
AP
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, but do not store above -20°C.
Pierce™
Biotin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.