missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Sulfo-SMCC (sulfosuccinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate), No-Weigh™ Format
Shop All Thermo Scientific Products
Click to view available options
Quantity:
50 mg
10 x 2 mg
1 g
Unit Size:
1g
50mg
Pack of 10
Description
Thermo Scientific™ Pierce Sulfo-SMCC is a very popular, water-soluble, amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a medium-length cyclohexane spacer arm (8.3 Ås).
Highlights:
- Water-soluble
- Reactive groups: sulfo-NHS ester and iodoacetyl
- Reactive toward: amino and sulfhydryl groups
Specifications
Specifications
| Chemical Reactivity | Amine-Sulfhydryl |
| Cleavable | No |
| Description | Sulfo-SMCC |
| Molecular Weight (g/mol) | 436.37 |
| PEGylated | No |
| Spacer Arm Length | 8.3 Å |
| Content And Storage | Upon receipt store desiccated at -20°C. |
| Cell Permeability | No |
| Shipping Condition | Ambient |
| Product Line | Pierce |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction