Learn More
Thermo Scientific™ Sulfo-SMCC (sulfosuccinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate), No-Weigh™ Format
Crosslink amines to sulfhydryls with this mid-length, water-soluble crosslinker made of NHS-ester and maleimide groups at ends of a cyclohexane-containing, 8-atom spacer arm.
£223.00 - £2390.00
Specifications
Chemical Reactivity | Amine-Sulfhydryl |
---|---|
Cleavable | No |
Description | Sulfo-SMCC |
Molecular Weight (g/mol) | 436.37 |
PEGylated | No |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15971227
|
Thermo Scientific™
A39268 |
10 x 2 mg |
£223.00
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13426629
|
Thermo Scientific™
22322 |
50 mg |
£311.00
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10698114
|
Thermo Scientific™
22122 |
1 g |
£2390.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thermo Scientific™ Pierce Sulfo-SMCC is a very popular, water-soluble, amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a medium-length cyclohexane spacer arm (8.3 Ås).
Highlights:
- Water-soluble
- Reactive groups: sulfo-NHS ester and iodoacetyl
- Reactive toward: amino and sulfhydryl groups
Specifications
Amine-Sulfhydryl | |
Sulfo-SMCC | |
No | |
Upon receipt store desiccated at -20°C. | |
Ambient | |
Heterobifunctional | |
Short (<10 Å) | |
Standard, Single-use, Premium-grade | |
Crosslinker |
No | |
436.37 | |
8.3 Å | |
No | |
Chemical Labeling | |
Maleimide, Sulfo-NHS Ester | |
Water | |
Yes |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.