Learn More
Thermo Scientific™ Pierce™ Fast Western Blot Kits, SuperSignal™ West Femto, Rabbit
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Pierce Fast Western Blot Kit, SuperSignal West Femto Substrate contains optimized reagents and protocol that allows mid to low femtogram levels of detection and shortens typical Western blot development to less than 1 hour after the gel transfer.
Highlights:
Fast – low femtogram levels of detection in about 1 hour
Convenient – no expensive hardware or vacuum required; no clogging issues
Simple – optimized protocol makes Western blot analysis easier than ever
Economical – no additional consumables or extra equipment to purchase
Stable – all kit components are stable for one year stored at 4°C
Easy – complete kits contain all components needed to block, probe and develop a blot with any standard mouse or rabbit primary antibody
Compatible with:
Pierce Membranes and Filter Paper; CL-Xposure™ Film for Western blotting; Pierce Western Blot Signal Enhancer; Restore Plus™ Western Blot Stripping Buffer
Specifications
Specifications
| Description | Pierce Fast Western Blot Kit, SuperSignal West Femto Substrate, Rabbit |
| Product Type | Western Blot Kit |
| Detection Method | Chemiluminescence |
| Form | Liquid |
| For Use With (Application) | Western Blot |
| Membrane Compatibility | Nitrocellulose, PVDF |
| Product Line | Pierce, SuperSignal |
| Quantity | 400 mL |
| Conjugate | HRP |
| Substrate | Horseradish Peroxidase |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.