missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ SuperSignal West Femto Fast Western Blot Kit

For mid-to-low femtogram-level detection and Western blot development less than 1 hour after gel transfer.

£116.00

Specifications

Detection Method Chemiluminescence
Quantity 80 mL Kit
Conjugate HRP
View More Specs

Products 1
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
10417764
View Documents
Thermo Scientific™
35086
Pierce Fast Western Blot Kit, SuperSignal West Femto Substrate, Rabbit Trial Size This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Thermo Scientific™ Pierce Fast Western Blot Kit, SuperSignal West Femto Substrate contains optimized reagents and protocol that allows mid to low femtogram levels of detection and shortens typical Western blot development to less than 1 hour after the gel transfer.

The Fast Western Blot Kit protocol is a quick and efficient method for developing Western blots without the need to purchase expensive instrumentation and consumables required by other rapid Western blotting systems. These complete Pierce Fast Western Blot Kits require minimal hands-on time and yield results comparable to classic Western blotting with SuperSignal West Femto Chemiluminescent Substrate. The kits include SuperSignal West Femto Substrate, the most sensitive enhanced chemiluminescent substrate available for detecting horseradish peroxidase (HRP). This substrates sensitivity, intensity and duration of the signal allow for easy documentation using photographic or other imaging methods. The developed blots can be repeatedly exposed and stripped of the primary antibody and reprobed.

Highlights:

  • Fast – low femtogram levels of detection in about 1 hour

  • Convenient – no expensive hardware or vacuum required; no clogging issues

  • Simple – optimized protocol makes Western blot analysis easier than ever

  • Economical – no additional consumables or extra equipment to purchase

  • Stable – all kit components are stable for one year stored at 4°C

  • Easy – complete kits contain all components needed to block, probe and develop a blot with any standard mouse or rabbit primary antibody

Compatible with:

Pierce Membranes and Filter Paper; CL-Xposure™ Film for Western blotting; Pierce Western Blot Signal Enhancer; Restore Plus™ Western Blot Stripping Buffer

Specifications

Specifications

Chemiluminescence
80 mL Kit
HRP
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.