Learn More
Thermo Scientific™ Pierce™ Fast Western Kit, SuperSignal™ West Pico, Rabbit
For picogram-level detection and Western blot development less than 1 hour after gel transfer.
£155.00 - £317.00
Specifications
Product Type | Western Blot Kit |
---|---|
Detection Method | Chemiluminescent |
Form | Liquid |
For Use With (Application) | Western Blot |
Membrane Compatibility | Nitrocellulose, PVDF |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10481945
|
Thermo Scientific™
35061 |
Pierce Fast Western Blot Kit, SuperSignal West Pico Substrate, Rabbit |
£317.00
1 set |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||||
10574914
|
Thermo Scientific™
35066 |
Pierce Fast Western Blot Kit, SuperSignal West Pico Substrate, Rabbit Trial Size |
£155.00
1 set |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
Description
Thermo Scientific™ Pierce Fast Western Blot Kit, SuperSignal West Pico Substrate contains optimized reagents and protocol that allows picogram levels of detection and shortens typical Western blot development to less than 1 hour after the gel transfer.
These Fast Western Blot Kits require minimal hands-on time and yield results comparable to classic Western blotting with SuperSignal West Pico Chemiluminescent Substrate.The kits contain all the reagents necessary to complete a Western blot being probed with either a mouse or rabbit primary antibody. Like other Western blotting methods, the included SuperSignal West Pico Substrate produces a chemiluminescent signal that is detectable using photographic or other imaging methods. The developed blots can be repeatedly exposed and stripped of the primary antibody and reprobed.Highlights:
Fast – picogram levels of detection in about one hour
Convenient – no expensive hardware or vacuum required; no clogging issues
Simple – optimized protocol makes Western blot analysis easier than ever
Economical – no additional consumables or extra equipment to purchase
Stable – all kit components are stable for one year stored at 4°C
Easy – complete kits contain all components needed to block, probe and develop a blot with any standard mouse or rabbit primary antibody
Compatible with:
Pierce Membranes and Filter Paper; CL-Xposure™ Film for Western blotting; Pierce Western Blot Signal Enhancer; Restore Plus™ Western Blot Stripping Buffer
Specifications
Western Blot Kit | |
Liquid | |
Nitrocellulose, PVDF | |
HRP | |
Rabbit |
Chemiluminescent | |
Western Blot | |
Pierce™, SuperSignal™ | |
Horseradish Peroxidase | |
Picogram-level |
For Research Use Only. Not for use in diagnostic procedures.