Learn More
Thermo Scientific™ TMEA (tris(2-maleimidoethyl)amine)
A 3-way, sulfhydryl-to-sulfhydryl crosslinker; tris (2-maleimidoethyl) amine; composed of three maleimide groups on short spacer arms; conjugate protein or peptide cysteines.
Brand:  Thermo Scientific™ 33043
Product Code. 11809250
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Thermo Scientific™ Pierce TMEA is a trifunctional crosslinker containing three reactive maleimide groups to enable three-way conjugation and polymerization of sulfhydryl-containing molecules.
Highlights:
  • Reactive group: maleimide
  • Reactive toward: sulfhydryl groups
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sulfhydryl-Sulfhydryl
TMEA
No
Upon receipt store at 4°C protected from moisture.
Ambient
Chemical Labeling
Maleimide
Solid
DMF, DMSO
No
No
386.36
10.3 Å
Yes
Pierce™
Homotrifunctional
Medium (10 to 30 Å)
50 mg
Standard
Crosslinker
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ TMEA (tris(2-maleimidoethyl)amine) > TMEA, For 3-way crosslinking between sulfhydryl molecules