missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Triethylamine (TEA), Sequencing Grade

Bottles of 99.5%-pure TEA (MW 101.2, CAS 121-44-8) liquid (sold by weight); for HPLC use as an ion-pairing agent in reverse-phase peptide separations.

Brand:  Thermo Scientific™ 25108

153.90 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.



Product Code. 11831385

  • £171.00 / 100g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce Triethylamine (TEA) is a high-purity solvent that can be used in certain HPLC protocols, such as weak anion exchange to resolve certain tryptic peptides on a reverse-phase column.

Triethylamine is an ion-pairing reagent that alters selectivity in reverse-phase HPLC separations. By pairing with peptides, it effectively sharpens peaks, resulting in improved peak resolution.

Highlights:

  • >99.5% triethylamine purity, allowing sensitive peptide detection at low UV wavelengths in reverse-phase HPLC peptide separation systems Packaged in amber glass bottles with protective PTFE-lined fluorocarbon caps for reagent integrity
  • Has a low UV absorbance to provide the most sensitive detection across all wavelengths

Properties of Triethylamine

  • Alternate names TEA, Diethylethanamine
  • Molecular formula C6H15N
  • Molecular weight 101.19
  • Density 0.726g/mL
  • CAS: 121-44-8

Ordering Information:

X in the ordering table indicates that hazardous shipping charges apply.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in original container protected from direct sunlight in a dry, cool and well-ventilated area.
Pierce™
100 g
Liquid
Ion Pairing Reagent
Triethylamine (TEA)
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138

    Expires2025-04-30

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Triethylamine (TEA), Sequencing Grade > 100g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.