missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pierce™ Triple Quadrupole Calibration Solution
Calibrate Triple Stage Quadrupole (TSQ) mass spectrometers using a mixture of high purity, ionizable components specifically designed for positive mode calibration.
Brand: Thermo Scientific™ 88325
This item is not returnable.
View return policy
Description
Thermo Scientific™ Pierce Triple Quadrupole Calibration Solution is a mixture of high purity, ionizable components specifically designed for positive mode calibration of Thermo Scientific™ Triple Stage Quadrupole instruments.
The Pierce Triple Quadrupole Calibration Solution is a ready-to-use liquid formulation ideal for quickly performing the required routine calibration to maintain the robust performance of Thermo Scientific Mass Spectrometers. The Triple Quadrupole Calibration Solution is manufactured at an ISO 9001 facility and each lot is quality controlled with strict specifications. The stable solution is provided in a leak-proof, high-purity PTFE bottle.Highlights
- Strong peaks — mixture of three tyrosine isotopes in methanol/formic acid solution
- Ready-to-use — load the reference standard into a syringe and inject into the instrument
- High-purity — mass spectrometry grade reagents in a a non-leachable container
- Stable — store at 2° to 8°C for more than one year
Specifications
Mass Spectrometer | |
Pierce™ | |
Small Molecules | |
Mass Spectrometry | |
Liquid |
Calibration Solution | |
Store between 2°C and 8°C. | |
Small Molecules | |
Calibration |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Pierce(R) Triple Quadrupole Calibration Solution, 10 mL
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Specific target organ toxicity Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H371-May cause damage to organs.
Precautionary Statement
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P330-Rinse mouth.
- P270-Do not eat, drink or smoke when using this product.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Methyl alcohol
For Research Use Only. Not for use in diagnostic procedures.