missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 3UV Lamp (230V, 50Hz)

Photoactivate crosslinkers in UV-crosslinking methods with these compact, UV-lamps with 252nm, 302nm and 365nm short to long wavelength bulbs.

Brand:  Thermo Scientific™ 95035

Product Code. 10531194

  • £720.00 / Each
Estimated Shipment: 31-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce UV Lamps are compact, multifunctional ultraviolet lamps with 252nm, 302nm and 365nm dial-selectable wavelength settings especially for use with photoactivatable crosslinkers and UV-crosslinking methods.

The 3UV lamp from UVP is a compact ultraviolet lamp for longwave, midrange and shortwave UV applications. The lamp's innovative design allows you to dial-in the required wavelength with just the turn of a knob. The lamp is compatible with our extensive line of photo-reactive crosslinking and label-transfer reagents, including the photo-reactive amino acids for discovering in-cell protein interactions.

Highlights:

  • Multifunctional – three lamps (254nm, 302nm, 365nm) selected by a turn of the dial

  • Efficient – unique reflector design provides maximum UV light for all applications

  • Lightweight and ergonomic – designed for hand-held or stationary use

  • Small footprint – 15.5 in. x 3 in. x 4.5 in. (394mm x 76mm x 114mm)

  • Durable – lamp features aluminum construction and scratch- resistant finish

  • Versatile – 8-watt lamps are ideal for a wide area of illumination, such as culture plates

  • Validated for crosslinking – our researchers have validated this lamp for use in crosslinking applications with diazirine and phenyl azide photoactivatable crosslinkers

TRUSTED_SUSTAINABILITY
Specifications

Specifications

UV
8 W
Pierce 3UV Lamp, 230 Volts, 50 Hz (Europe)
1 unit
230 V
365/302/254 nm
Triple Wavelength UV Lamp
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZJ4418491N Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number95035
Lot NumberZJ4418491I Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number95035
Lot NumberZJ4418491L Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number95035
Lot NumberZJ4418491X Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number95035
Lot NumberZJ4418491V Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number95035
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ 3UV Lamp (230V, 50Hz) > 230V, 50Hz

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.