Learn More
Thermo Scientific™ Vacuum Hydrolysis Tube, 1 mL
High-quality valved glass tubes designed for heated, vacuum, acid-hydrolysis of proteins and peptides for amino acid analysis by HPLC chromatography.
Brand: Thermo Scientific™ 29570
Code : PN
Additional Details : Weight : 0.01000kg
Description
Thermo Scientific™ Pierce Vacuum Hydrolysis Tubes are high-quality glass tubes designed for heated, acid-based, vacuum hydrolysis of proteins and peptides for amino acid analysis.
Highlights:
- Upper temperature limit of the Vacuum Hydrolysis Tubes is 260°C; however, do not heat the tubes greater than 110°C in an oven Vacuum hydrolysis tubes fit conveniently into Reacti-Block™ Aluminum Heating Blocks
Specifications
Vacuum Hydrolysis Tube, 1mL | |
0.0 | |
1 Tube |
Vacuum Hydrolysis Tube | |
Tube |
For Research Use Only. Not for use in diagnostic procedures.