missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Vacuum Hydrolysis Tube, 6 mL
High-quality valved glass tubes designed for heated, vacuum, acid-hydrolysis of proteins and peptides for amino acid analysis by HPLC chromatography.
Brand: Thermo Scientific™ 29571
This item is not returnable.
View return policy
Description
Thermo Scientific™ Pierce Vacuum Hydrolysis Tubes are high-quality glass tubes designed for heated, acid-based, vacuum hydrolysis of proteins and peptides for amino acid analysis.
Highlights:
- Upper temperature limit of the Vacuum Hydrolysis Tubes is 260°C; however, do not heat the tubes greater than 110°C in an oven Vacuum hydrolysis tubes fit conveniently into Reacti-Block™ Aluminum Heating Blocks
Specifications
Vacuum Hydrolysis Tube, 6mL | |
0.0 | |
1 Tube |
Vacuum Hydrolysis Tube | |
Tube |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction