Learn More
Thermo Scientific™ Water, LC-MS Grade
Thermo Scientific™ Water, LC/MS Grade is ultapure water for formulation of solvents for HPLC
Brand: Thermo Scientific™ 85189
Code : PN
Additional Details : CAS Number : 7732-18-5 Weight : 1.36080kg
Description
Thermo Scientific™ Water, LC/MS Grade is ultapure water for formulation of solvents for HPLC.
An ultrapure, LC/MS-grade preparation with low UV-absorptivity that makes it suitable and reliable for use in HPLC and mass spectrometry applications.
Thermo Scientific Water, LC/MS Grade is specially purified by a proprietary method and treated to ensure lot-to-lot consistency with a low UV absorbance to provide you with most sensitive detection across all wavelengths. Pierce Water is 0.2-micron filtered, packaged in solvent-rinsed amber glass bottles and sealed under a nitrogen atmoshere with Teflon TFE-lined fluorocarbon caps for ultimate protection.
Highlights:
- Purity and quality validated with 30 tests for use in LC/MS applications
- 0.2-micron filtered, packed in solvent-rinsed amber glass bottles
- Sealed under nitrogen atmosphere to eliminate absorption of unknown atmospheric gases
Product Details:
General properties of water
- Molecular formula: H2O
- Molecular Weight: 18.01
- CAS number: 7732-18-5
Water, LC/MS Grade is specifically purified and tested to the highest specifications to ensure the integrity of your data, maximize sensitivity in your assay and prolong the life of your equipment.
Specifications
7 | |
32°F | |
LC-MS | |
Liquid |
Clear, Colorless | |
212°F | |
4 x 1 L | |
Pierce™ |
For Research Use Only. Not for use in diagnostic procedures.