missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Western Blot Signal Enhancer

Product Code. 11836804 Shop All Thermo Scientific Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 11836804

Brand: Thermo Scientific™ 21050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Greatly enhance the effectiveness of primary antibodies and intensify the final Western blot detection signal with this two-reagent blot conditioning system.

Thermo Scientific™ Pierce Western Blot Signal Enhancer is a two-reagent system for conditioning protein blots after transfer to greatly enhance the effectiveness of primary antibodies and intensify the final detection signal in Western blot experiments.

The Pierce Western Blot Signal Enhancer membrane treatment procedure is very simple, takes only 15 minutes and can be added to nearly any existing Western blotting protocol. The result is an increase in the intensity of target protein bands on the Western blot or detection of target proteins at levels that were previously not possible. The product is effective for signal intensification with both chemiluminescent and chromogenic substrates, especially with nitrocellulose membranes.

Highlights:

  • Increases protein detection – most protein targets show a three- to 10-fold increase in signal intensity, enabling much less protein to be detected with the same substrate and method

  • Improves antibody binding – the membrane-treatment reagent exposes and conditions target proteins so that specific antibodies can bind more effectively

  • Works for nearly any protein – signal enhancement has been demonstrated with targets such as IL-6, p53, NFkB, BRCA1 and EGF

  • Effective with any substrate – enhances both chemiluminescent (ECL) and colorimetric detection for Western blots

  • Compatible with any membrane – enhances signal on nitrocellulose and PVDF membrane, regardless of pore size (enhancement is less pronounced with PVDF)

  • Fast, 15-minute protocol – optimized for a combination of simplicity, speed and signal enhancement for most proteins

  • Ready-to-use – no formulating or diluting necessary, and the reagents are stable for storage at room temperature

Includes:

Enhancer Reagent 1, 250mL; Enhancer Reagent 2, 250mL

TRUSTED_SUSTAINABILITY

Specifications

Product Type Western Blot Enhancer
Detection Method Colorimetric, Chemiluminescence
Form Liquid
For Use With (Application) Western Blot
Membrane Compatibility Nitrocellulose, PVDF
Product Line Pierce
Quantity 500 mL
Content And Storage Upon receipt store kit at room temperature.

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.