missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pig anti-Human IgG Fc Secondary Antibody, Novus Biologicals™
Pig Polyclonal Antibody
£223.00
Specifications
Dilution | ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoblotting 1:100 - 1:2000, Dot Blot 1:100-1:2000 |
---|---|
Applications | ELISA, Immunocytochemistry, Immunofluorescence, Immunoblot, Dot Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Pig |
Description
IgG Fc Polyclonal specifically detects IgG Fc in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunoblotting, Dot Blot.Specifications
ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoblotting 1:100 - 1:2000, Dot Blot 1:100-1:2000 | |
Polyclonal | |
Pig | |
Secondary |
ELISA, Immunocytochemistry, Immunofluorescence, Immunoblot, Dot Blot | |
Unconjugated | |
Human | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Pig anti-Human IgG Fc Secondary Antibody, Novus Biologicals™