missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pig anti-Human IgM Fc Secondary Antibody, Novus Biologicals™
Pig Polyclonal Antibody
£218.14
Specifications
Concentration | LYOPH |
---|---|
Dilution | Immunoprecipitation 1:10 - 1:500 |
Applications | Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
IgM Fc Polyclonal specifically detects IgM Fc in Human samples. It is validated for Immunoprecipitation.Specifications
LYOPH | |
Immunoprecipitation | |
Unconjugated | |
Porcine | |
Secondary |
Immunoprecipitation 1:10 - 1:500 | |
Polyclonal | |
Lyophilized | |
Human | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |