Learn More
Thermo Scientific™ Piko PCR Plate, 24-well, white
Thermo Scientific™ Piko PCR Plates are modular, ultra-thin wall PCR plates for Piko* Thermal Cyclers.
Brand: Thermo Scientific™ SPL024150
51.98 GBP valid until 2024-09-27
Use promo code "21650" to get your promotional price.
This item is not returnable.
View return policy
Description
Thermo Scientific™ Piko™ PCR Plates are modular, ultra-thin wall PCR plates for Piko Thermal Cyclers.
Features
• Ultra Thin Wall (UTW) for fast PCR/qPCR applications
• Low profile
• Designed for use with the Piko and PikoReal 24-well thermal cyclers
• Plates can be snapped into the plate frame to create a standard 96-well plate
• Compatible with standard multi-channel pipettes and liquid-handling platforms
• Well spacing and footprint conform to industry (ANSI) dimensions
Applications
• qPCR/Real-Time PCR
Related Products
Piko PCR Plate, 24-well, clear
Piko PCR Plate, 24-well, white
Plate Frame for 24-well Piko PCR Plates, white
Specifications
No Barcode | |
200 Plate | |
White | |
Piko Thermal Cycler, PikoReal Real-Time PCR System | |
24 |
Certified RNase/DNase-free | |
Piko | |
PCR Microplate | |
Polypropylene | |
Low profile |