Learn More
Thermo Scientific™ Piko PCR Plate, 96-well, white
Speed up PCR and qPCR applications with Piko 24-well PCR Plates, with ultra-thin wall (UTW) technology to dramatically reduce the thickness of the well walls.
Brand: Thermo Scientific™ SPL096150
46.08 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Piko PCR Plates are a family of PCR plates which have the footprint of a microscope slide. We have not only shrunk the size of the plate, but dramatically reduced the thickness of the well walls utilizing our patented ultra-thin wall (UTW) technology.
Four plates can be snapped into a single frame producing the equivalent of a standard 384-well microplate.Features
• Ultra Thin Wall (UTW) for fast PCR/qPCR applications
• Low profile
• Designed for use with the Piko and PikoReal 96-well thermal cyclers
• Plates can be snapped into the plate frame to create a standard 384-well plate
• Compatible with standard multi-channel pipettes and liquid-handling platforms
• Well spacing and footprint conform to industry (ANSI) dimensions
Applications
• qPCR/Real-Time PCR
Related Products
Piko PCR Plate, 96-well, clear
Piko PCR Plate, 96-well, white
Plate Frame for 96-well Piko PCR Plates, white
Specifications
No Barcode | |
50 Plate | |
White | |
Piko Thermal Cycler, PikoReal Real-Time PCR System | |
96 |
Certified RNase/DNase-free | |
Piko | |
PCR Microplate | |
Polypropylene | |
Standard |
For Research Use Only. Not for use in diagnostic procedures.