missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Pimecrolimus
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mg
500 mg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | White to beige |
Purity | ≥96.0% by HPLC |
Packaging | Glass Bottle |
Content And Storage | Refrigerator +4°C |
Shipping Condition | normal conditions |
Source | synthetic source |
Quantity | 500 mg |
Product Type | Pimecrolimus |
Molecular Weight (g/mol) | 810.45 |
Form | Powder |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'