missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pimobendan, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
Concentration | 97.5% to 102.5% |
Packaging | Glass Bottle |
Quantity | 250 mg |
Color | White to Light Yellow |
Certificate of Analysis | Available |
Concentration or Composition Notes | Titration with HClO4 |
UN Number | 2811 |
Merck Index | 7516 |
Formula Weight | 334.37 |
Physical Form | Powder |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'