missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIN4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA544596
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Peptide sequence: MPMAGLLKGL VRQLERFSVQ QQASKMPPKG KSGSGKAGKG GAASGSDSAD Sequence homology: Human: 100%; Rat: 90%
PIN4 Polyclonal antibody specifically detects PIN4 in Human samples. It is validated for Western Blot.Spezifikation
PIN4 | |
Polyclonal | |
Unconjugated | |
Pin4 | |
2410002I22Rik; EPVH; eukaryotic parvulin homolog; hEPVH; hPar14; hPar17; MGC138486; Par14; Par17; parvulin; parvulin-14; Parvulin-17; peptidyl-prolyl cis/trans isomerase EPVH; peptidylprolyl cis/trans isomerase NIMA-interacting, 4; peptidylprolyl cis/trans isomerase, NIMA-interacting 4; peptidyl-prolyl cis-trans isomerase NIMA-interacting 4; Peptidyl-prolyl cis-trans isomerase Pin4; PIN4; PPIase Pin4; protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting, 4 (parvulin); protein (peptidylprolyl cis/trans isomerase) NIMA-interacting, 4 (parvulin); rotamase PIN4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
5303 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9Y237 | |
Pin4 | |
Synthetic peptide directed towards the N-terminal of human PIN4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |