Learn More
Ohaus™ Pioneer™ PX 224 Analytical Balance
The Pioneer PX combines essential weighing functionality with competitive performance, offering high accuracy and repeatability for applications in laboratory, industrial and education settings.
Brand:  Ohaus™ 30429804
1250.62 GBP valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Product Code. 15875327
  • £2095.00 / Each
Explore more special offers
Description
Description
  • Competitive Performance at an Economical Price - The PX offers high accuracy and repeatability for essential weighing applications in laboratory, industrial and educational settings at an economical pricepoint.
  • Durably Constructed for Long-Term Use - Featuring a cast metal lower housing, sub-pan and stainless steel weighing pan, the PX is durably constructed for versatile, long-term use.
  • Designed for Simple, Intelligent Operation - The PX features a second line display for additional information or guidance, a static removal bar for convenient grounding, and USB and RS232 connectivity for easy communication.
TRUSTED_SUSTAINABILITY
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Ohaus™ Pioneer™ PX 224 Analytical Balance > Capacity: 220 g