Learn More
Marigold™ G26G Green Nitrile™ Chemical Resistant Gloves
12.91 GBP valid until 2024-07-31
Use promo code "21640" to get your promotional price.
Use promo code "21640" to get your promotional price.
Click to view available options
Size: X-Large
Description
- Chemical and liquid protection gloves
- Good abrasion resistance
- Flexible and comfortable for good dexterity
- Superior raised grip pattern for safety handling
- Food approved
- Cotton liner
- Single chlorination
Specifications
Color | Green |
Material | Nitrile |
Size | X-Large |