missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Pipe Markers (On Liner) - Standard Legends
European Standard Pipemarkers reinforce safety and aid maintenance.
£12.30 - £15.25
Specifications
Certifications/Compliance | ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379 |
---|---|
Description | Individual Pipe Markers on a Card with die-cut arrowheads, without pictograms - Acids and Alkalis - Acid |
Adhesive Type | Permanent Acrylic, Permanent, Pressure-sensitive |
Color | White on Purple |
Finish | Gloss |
Product Code | Brand | Width (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) | Price | Quantity & Availability | |||||
15339015
|
Brady™
N006987 |
450 mm |
£12.30
Pack of 2 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15309015
|
Brady™
N006985 |
250 mm |
£15.25
Pack of 4 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
Description
- Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
- Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, which saves you having to identify the appropriate symbol.
- Markers are printed with a contrasting border in the same color as the text for improved visibility.
- Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required
Specifications
ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379 | |
Permanent Acrylic, Permanent, Pressure-sensitive | |
Gloss | |
Pipe Marker | |
Pipe and valve marking | |
-40°F to +230°F | |
0.0039 in. |
Individual Pipe Markers on a Card with die-cut arrowheads, without pictograms - Acids and Alkalis - Acid | |
White on Purple | |
Polyester | |
Acid | |
English | |
-40°C to +110°C | |
0.099 mm |