missing translation for 'onlineSavingsMsg'
Learn More

Piperazine, 99%, extra pure, Thermo Scientific Chemicals

Product Code. 10115493
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-85-0
Molecular Formula C4H10N2
Molecular Weight (g/mol) 86.14
MDL Number MFCD00005953
InChI Key GLUUGHFHXGJENI-UHFFFAOYSA-N
Synonym diethylenediamine, piperazin, 1,4-diazacyclohexane, hexahydropyrazine, piperazidine, antiren, 1,4-piperazine, diethyleneimine, eraverm, pipersol
PubChem CID 4837
ChEBI CHEBI:28568
IUPAC Name piperazine
SMILES C1CNCCN1

Specifications

Melting Point 107.0°C to 111.0°C
Color White to Yellow
Boiling Point 146.0°C to 148.0°C
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic drum
Quantity 2.5 kg
Beilstein 23,4
Fieser 04,392
Merck Index 15,7576
Solubility Information Solubility in water: 150g/L (20°C). Other solubilities: freely soluble in glycerol and glycols, soluble in alcohol, practically insoluble in diethyl ether
Formula Weight 86.14
Percent Purity 99%
Grade Extra Pure
Physical Form Crystalline Flakes
Chemical Name or Material Piperazine, anhydrous, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'