missing translation for 'onlineSavingsMsg'
Learn More

Piperazine, 99%, extra pure, Thermo Scientific Chemicals

Product Code. 10785611
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-85-0
Molecular Formula C4H10N2
Molecular Weight (g/mol) 86.14
MDL Number MFCD00005953
InChI Key GLUUGHFHXGJENI-UHFFFAOYSA-N
Synonym diethylenediamine, piperazin, 1,4-diazacyclohexane, hexahydropyrazine, piperazidine, antiren, 1,4-piperazine, diethyleneimine, eraverm, pipersol
PubChem CID 4837
ChEBI CHEBI:28568
IUPAC Name piperazine
SMILES C1CNCCN1

Specifications

Melting Point 107.0°C to 111.0°C
Color White to Yellow
Boiling Point 146.0°C to 148.0°C
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Quantity 100 g
Beilstein 23,4
Fieser 04,392
Merck Index 15,7576
Solubility Information Solubility in water: 150g/L (20°C). Other solubilities: freely soluble in glycerol and glycols, soluble in alcohol, practically insoluble in diethyl ether
Formula Weight 86.14
Percent Purity 99%
Grade Extra Pure
Physical Form Crystalline Flakes
Chemical Name or Material Piperazine, anhydrous, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Piperazine
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Reproductive toxicity Category 2
Hazard Statement
  • H228-Flammable solid.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'