Learn More
Piperazine, anhydrous, 99%, Thermo Scientific Chemicals
£20.90 - £159.80
Chemical Identifiers
CAS | 110-85-0 |
---|---|
Molecular Formula | C4H10N2 |
Molecular Weight (g/mol) | 86.14 |
MDL Number | MFCD00005953 |
InChI Key | GLUUGHFHXGJENI-UHFFFAOYSA-N |
Synonym | diethylenediamine, piperazin, 1,4-diazacyclohexane, hexahydropyrazine, piperazidine, antiren, 1,4-piperazine, diethyleneimine, eraverm, pipersol |
PubChem CID | 4837 |
ChEBI | CHEBI:28568 |
IUPAC Name | piperazine |
SMILES | C1CNCCN1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11447317
|
Thermo Scientific Alfa Aesar
A15049.22 |
100 g |
£20.90
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11457317
|
Thermo Scientific Alfa Aesar
A15049.36 |
500 g |
£47.24
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11467317
|
Thermo Scientific Alfa Aesar
A15049.0E |
2.5 kg |
£159.80
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Piperazine acts as an intermediate for pharmaceuticals, polymers, dyes, corrosion inhibitors, rubber accelerators and surfactants. It is used as a raw material in the manufacture of plastics, polyamide resins, urethane and brake fluid. It is also used for carbondioxide and hydrogen sulfide gas scrubbing mediums. Furthermore, it serves as an epoxy curing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPiperazine acts as an intermediate for pharmaceuticals, polymers, dyes, corrosion inhibitors, rubber accelerators and surfactants. It is used as a raw material in the manufacture of plastics, polyamide resins, urethane and brake fluid. It is also used for carbondioxide and hydrogen sulfide gas scrubbing mediums. Furthermore, it serves as an epoxy curing agent.
Solubility
Soluble in water, chloroform, glycerol, glycols and alcohol. Slightly soluble in benzene and heptane. Insoluble in ether.
Notes
Air and light sensitive. Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong acids.
Chemical Identifiers
110-85-0 | |
86.14 | |
GLUUGHFHXGJENI-UHFFFAOYSA-N | |
4837 | |
piperazine |
C4H10N2 | |
MFCD00005953 | |
diethylenediamine, piperazin, 1,4-diazacyclohexane, hexahydropyrazine, piperazidine, antiren, 1,4-piperazine, diethyleneimine, eraverm, pipersol | |
CHEBI:28568 | |
C1CNCCN1 |
Specifications
110-85-0 | |
1.11 | |
88°C (190°F) | |
MFCD00005953 | |
102555 | |
14,7464 | |
Soluble in water,chloroform,glycerol,glycols and alcohol. Slightly soluble in benzene and heptane. Insoluble in ether. | |
C1CNCCN1 | |
86.14 | |
CHEBI:28568 | |
99% |
108°C to 113°C | |
146°C to 148°C | |
C4H10N2 | |
UN2579 | |
Hygroscopic | |
diethylenediamine, piperazin, 1,4-diazacyclohexane, hexahydropyrazine, piperazidine, antiren, 1,4-piperazine, diethyleneimine, eraverm, pipersol | |
GLUUGHFHXGJENI-UHFFFAOYSA-N | |
piperazine | |
4837 | |
86.14 | |
Piperazine, anhydrous |
Safety and Handling
GHS H Statement
H334-H361-H314-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of damaging fertility or the unborn child.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
P201-P202-P210-P240-P241-P260-P264b-P272-P280g-P281-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P370+P378q-P501c
H228-H314-H317-H334-H361fd
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: PIPERAZINE
missing translation for 'einecsNumber' : 203-808-3
missing translation for 'rtecsNumber' : TK7800000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Piperazine, anhydrous
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 1
- Reproductive toxicity Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
Hazard Statement
- H228-Flammable solid.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : Piperazine
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.