Learn More
Piperazine hydrate, ≥97%, Thermo Scientific Chemicals
£35.30 - £309.00
Chemical Identifiers
CAS | 142-63-2 |
---|---|
Molecular Formula | C4H22N2O6 |
Molecular Weight (g/mol) | 194.228 |
MDL Number | MFCD00149389 |
InChI Key | AVRVZRUEXIEGMP-UHFFFAOYSA-N |
Synonym | piperazine hexahydrate, arthriticine, piperazine, hexahydrate, vermisol, parid, unii-p3m07b8u64, usaf a-3803, diethylenediamine hexahydrate, vermyl tn |
PubChem CID | 120181 |
IUPAC Name | piperazine;hexahydrate |
SMILES | C1CNCCN1.O.O.O.O.O.O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15437737
|
Thermo Scientific Alfa Aesar
H37069.30 |
250 g |
£35.30
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15417737
|
Thermo Scientific Alfa Aesar
H37069.0B |
1000 g |
£79.90
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15427737
|
Thermo Scientific Alfa Aesar
H37069.0I |
5000 g |
£309.00
5000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
142-63-2 | |
194.228 | |
AVRVZRUEXIEGMP-UHFFFAOYSA-N | |
120181 | |
C1CNCCN1.O.O.O.O.O.O |
C4H22N2O6 | |
MFCD00149389 | |
piperazine hexahydrate, arthriticine, piperazine, hexahydrate, vermisol, parid, unii-p3m07b8u64, usaf a-3803, diethylenediamine hexahydrate, vermyl tn | |
piperazine;hexahydrate |
Specifications
142-63-2 | |
White | |
125°C to 130°C | |
Weak, characteristic | |
MFCD00149389 | |
piperazine hexahydrate, arthriticine, piperazine, hexahydrate, vermisol, parid, unii-p3m07b8u64, usaf a-3803, diethylenediamine hexahydrate, vermyl tn | |
C1CNCCN1.O.O.O.O.O.O | |
194.228 | |
86.14 (Anhydrous) | |
Crystalline Powder or Flakes |
42°C to 46°C | |
11 to 12 | |
>100°C (212°F) | |
C4H22N2O6 | |
UN2579 | |
AVRVZRUEXIEGMP-UHFFFAOYSA-N | |
piperazine;hexahydrate | |
120181 | |
≥97% | |
Piperazine hydrate |
Safety and Handling
GHS H Statement
H334-H361-H314-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of damaging fertility or the unborn child.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P271-P272-P280g-P281-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
H314-H317-H334-H335-H361fd
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: PIPERAZINE
missing translation for 'einecsNumber' : 203-808-3
missing translation for 'rtecsNumber' : TM0850000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only