missing translation for 'onlineSavingsMsg'
Learn More
Learn More
piperidine-1-carboximidamide hydroiodide, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Assay Percent Range | 97% |
Quantity | 1 g |
Formula Weight | 255.1 |
Percent Purity | 97% |
Chemical Name or Material | Piperidine-1-carboximidamide hydroiodide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'