missing translation for 'onlineSavingsMsg'
Learn More

Piperidine, Pure, Fisher Chemical™

Catalog Number 10558630
Click to view available options
Quantity:
100 mL
250 mL
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
110-89-4
C5H11N
85.15
5979
NQRYJNQNLNOLGT-UHFFFAOYSA-N
hexahydropyridine, azacyclohexane, cyclopentimine, cypentil, hexazane, pentamethyleneimine, piperidin, pentamethylenimine, perhydropyridine, pentamethyleneamine
8082
CHEBI:18049
piperidine
C1CCNCC1
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
110-89-4
C5H11N
85.15
5979
NQRYJNQNLNOLGT-UHFFFAOYSA-N
hexahydropyridine, azacyclohexane, cyclopentimine, cypentil, hexazane, pentamethyleneimine, piperidin, pentamethylenimine, perhydropyridine, pentamethyleneamine
8082
CHEBI:18049
piperidine
C1CCNCC1
Color Colorless
Boiling Point 106°C
Melting Point -11°C
Formula Weight 85.15g/mol
Packaging Plastic Coated Glass bottle
Viscosity 1.46 mPaS at 20°C
Vapor Pressure 24.75mmHg at 20°C
Vapor Density 3
pH 12.6
Quantity 100 mL
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Piperidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'