missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piperine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 128.0°C to 132.0°C |
Color | Tan to White or Tan-Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (HPLC) |
Packaging | Glass bottle |
Quantity | 5 g |
Merck Index | 15, 7584 |
Solubility Information | Solubility in water: 40 mg/l (18°C). Other solubilities: 1g/15mL alcohol - 1g/1.7mL chloroform, 1g/36mL ether, soluble in benzene and acetic acid, practically insoluble in petroleum ether |
Formula Weight | 285.34 |
Percent Purity | 98% |
Show More |
Safety and Handling
Product Identifier
- Piperine
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'