missing translation for 'onlineSavingsMsg'
Learn More

Piperonyl alcohol, 98%, Thermo Scientific Chemicals

Product Code. 10499440
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 495-76-1
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00005836
InChI Key BHUIUXNAPJIDOG-UHFFFAOYSA-N
Synonym piperonyl alcohol, piperonol, 1,3-benzodioxole-5-methanol, 3,4-methylenedioxybenzyl alcohol, benzo d 1,3 dioxol-5-ylmethanol, piperonylalcohol, 5-hydroxymethyl-1,3-benzodioxole, 1-hydroxymethyl-3,4-methylenedioxybenzene, benzo 1,3 dioxol-5-ylmethanol, 3,4-methylenedioxy phenylmethanol
PubChem CID 10322
IUPAC Name 1,3-benzodioxol-5-ylmethanol
SMILES C1OC2=C(O1)C=C(C=C2)CO

Specifications

Melting Point 51.0°C to 55.0°C
Color White
Boiling Point 156.0°C to 158.0°C (66.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 100 g
Beilstein 19, 67
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol, diethyl ether, benzene,, soluble in chloroform
Formula Weight 152.15
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Piperonyl alcohol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'