Learn More
Thermo Scientific Chemicals PIPES, 0.5M buffer soln., pH 7.0
£65.30 - £101.00
Chemical Identifiers
CAS | 5625-37-6 |
---|---|
Molecular Formula | C8H18N2O6S2 |
Molecular Weight (g/mol) | 302.36 |
MDL Number | MFCD00006159 |
InChI Key | IHPYMWDTONKSCO-UHFFFAOYSA-N |
PubChem CID | 79723 |
ChEBI | CHEBI:44933 |
IUPAC Name | 2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid |
SMILES | C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15495489
|
Thermo Scientific Chemicals
J63234.AE |
100 mL |
£65.30
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15405499
|
Thermo Scientific Chemicals
J63234.AK |
250 mL |
£101.00
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PIPES, is used as a buffering agent in biochemistry. PIPES has pKa near the physiological pH which makes it useful in cell culture work. PIPES has been documented minimizing lipid loss when buffering glutaraldehyde histology in plant and animal tissues.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPIPES, is used as a buffering agent in biochemistry. PIPES has pKa near the physiological pH which makes it useful in cell culture work. PIPES has been documented minimizing lipid loss when buffering glutaraldehyde histology in plant and animal tissues.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Chemical Identifiers
5625-37-6 | |
302.36 | |
IHPYMWDTONKSCO-UHFFFAOYSA-N | |
CHEBI:44933 | |
C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O |
C8H18N2O6S2 | |
MFCD00006159 | |
79723 | |
2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid |
Specifications
5625-37-6 | |
0.5 M | |
Liquid | |
MFCD00006159 | |
IHPYMWDTONKSCO-UHFFFAOYSA-N | |
2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid | |
79723 | |
PIPES buffer soln. |
Colorless | |
7.0 | |
C8H18N2O6S2 | |
Soluble in water. | |
C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O | |
302.36 | |
CHEBI:44933 |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only