missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIPES, 1.0M buffer soln., pH 6.0, Thermo Scientific™
£57.06 - £107.00
Chemical Identifiers
CAS | 5625-37-6 |
---|---|
Molecular Formula | C8H18N2O6S2 |
Molecular Weight (g/mol) | 302.36 |
MDL Number | MFCD00006159 |
InChI Key | IHPYMWDTONKSCO-UHFFFAOYSA-N |
PubChem CID | 79723 |
ChEBI | CHEBI:44933 |
IUPAC Name | 2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid |
SMILES | C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15413359
|
Thermo Scientific Alfa Aesar
J60611.AK |
250 mL | |||||||
15423359
|
Thermo Scientific Alfa Aesar
J60611.AP |
500 mL |
£107.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
5625-37-6 | |
302.36 | |
IHPYMWDTONKSCO-UHFFFAOYSA-N | |
CHEBI:44933 | |
C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O |
C8H18N2O6S2 | |
MFCD00006159 | |
79723 | |
2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid |
Specifications
5625-37-6 | |
1.0 M | |
Liquid | |
MFCD00006159 | |
C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O | |
302.36 | |
CHEBI:44933 |
Colorless | |
6.0 | |
C8H18N2O6S2 | |
IHPYMWDTONKSCO-UHFFFAOYSA-N | |
2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid | |
79723 | |
PIPES buffer soln. |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold