missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIPES Sesquisodium Salt, MP Biomedicals™
185.16 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Quantity:
100 g
Description
- PIPES is frequently used as a buffering agent in biochemistry.
- It is an ethanesulfonic acid buffer developed by Good et al. in the 1960s.
- PIPES has a pKa near the physiological pH which makes it useful in cell culture work.
Specifications
Color | White |
Physical Form | Crystalline Powder |
Quantity | 100 g |
Assay Percent Range | 1.001 |
SMILES | C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)[O-].C1CN(CCN1CCS(=O)(=O)[O-])CCS(=O)(=O)[O-].[Na+].[Na+].[Na+] |
IUPAC Name | trisodium;2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonate;2-[4-(2-sulfonatoethyl)piperazin-1-yl]ethanesulfonate |
Molecular Weight (g/mol) | 670.665 |
Formula Weight | 335.3 |
Chemical Name or Material | PIPES |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'