missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIPES, Sesquisodium Sodium Salt, Fisher BioReagents
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Description
This biological buffer has a usable pH range of 6.1 to 7.5.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 300°C |
Color | White |
pH | 6.5 to 7.1 |
Physical Form | Powder or Solid |
Quantity | 500 g |
DNase | DNase free |
Assay Percent Range | ≥95 % |
Identification | Pass Test |
Packaging | Poly Bottle |
Protease | Protease free |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'