missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Pipet Stands and Accessories

Optimize the use of mLINE™ and Proline™ mechanical pipets. Sartorius Pipet Stands and Accessories include single- and multi-pipet stands, colored-coded caps and a calibration tool. Can also be used with a range of standard pipet brands.

£22.24 - £121.00

Specifications

For Use With (Equipment) mLINE™/Proline™ Plus mechanical pipette
View More Specs

Products 2
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
10398222
View Documents Promotion Details
Sartorius
725620
Linear Pipet Stand
£121.00
Each
In Stock
to see stock.
Edge
Add to basket
 
15514029
View Documents
Sartorius
726001
Color Coded Caps This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Pipette Stands
  • Stores pipettes in an upright position while not in use to avoid contamination from the desktop
    • Linear Stand is designed for all mechanical devices and stores them neatly in a line for instant access
    • Carousel stand is ideal for saving bench space and stores up to 6 mechanical pipettes
    • mLINE/Proline Plus Single Holder compactly holds a single mechanical pipette
Color Coded Caps
  • Colored caps help identify the volume of mLINE pipettes so it can easily be matched to the corresponding tip size
  • Includes 5 color-coded replacement caps
Calibration Tool
  • Calibration Tool for mLINE or Proline Plus mechanical pipettes for continuous accuracy and precision
  • Can also be used as a tube opener
Specifications

Specifications

mLINE™/Proline™ Plus mechanical pipette
Videos
SDS
Documents

Documents

Product Certifications